Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanedioic acid, 1-(1,1-dimethylethyl) ester | 234081-96-0 | MFCD28505598 | 98.0% | 258.35 g·mol⁻1 | C14H26O4 | 1 ML
SDP

Catalog No. 5000300534
Encompass_Preferred

10-(tert-Butoxy)-10-oxodecanoic acid is a short organic linker containing a terminal carboxylic acid and a tert-butyl ester protecting group, commonly used in organic synthesis and bioconjugation. It is supplied as a solid or as a 10 mM solution in DMSO for assay use, and is formulated for straightforward incorporation into conjugation workflows.

  • Contains carboxylic acid and tert-butyl ester functional groups.
  • Useful as a protected carboxylic acid linker for conjugation chemistry.
  • Available as 10 mM solution in DMSO or as bulk solid for synthesis.
  • Colorless to light yellow, solid-liquid mixture.
  • Molecular formula C14H26O4 and molecular weight 258.35 g·mol⁻1.
  • High purity (98.0%) suitable for research applications.

Catalog No. 50-003-00534 Supplier Medchemexpress LLC Supplier No. HYW03945810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

10-(tert-Butoxy)-10-oxodecanoic acid is a short organic linker containing a terminal carboxylic acid and a tert-butyl ester protecting group, commonly used in organic synthesis and bioconjugation. It is supplied as a solid or as a 10 mM solution in DMSO for assay use, and is formulated for straightforward incorporation into conjugation workflows.

  • Contains carboxylic acid and tert-butyl ester functional groups.
  • Useful as a protected carboxylic acid linker for conjugation chemistry.
  • Available as 10 mM solution in DMSO or as bulk solid for synthesis.
  • Colorless to light yellow, solid-liquid mixture.
  • Molecular formula C14H26O4 and molecular weight 258.35 g·mol⁻1.
  • High purity (98.0%) suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.