Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanedioic acid, 1,10-bis(2,2,6,6-tetramethyl-4-piperidinyl) ester | 52829-07-9 | 98.0% | 480.73 | 1 KG
SDP

Catalog No. 5000454241
Encompass_Preferred

BTMPS is a nicotine receptor (nAChR) antagonist that can lessen the negative effects of morphine on rats. It is for research use only and not sold to patients.

  • Appearance is solid
  • Color is white to off-white
  • Shipping is at room temperature in continental US; may vary elsewhere
  • Storage is at 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility (in vitro): DMSO: 10 mg/mL (20.80 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO).

Catalog No. 50-004-54241 Supplier Medchemexpress LLC Supplier No. HYW0131131KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BTMPS is a nicotine receptor (nAChR) antagonist that can lessen the negative effects of morphine on rats. It is for research use only and not sold to patients.

  • Appearance is solid
  • Color is white to off-white
  • Shipping is at room temperature in continental US; may vary elsewhere
  • Storage is at 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Solubility (in vitro): DMSO: 10 mg/mL (20.80 mM; ultrasonic and warming and heat to 60°C; Hygroscopic DMSO has a significant impact on the solubility of product, please use newly opened DMSO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.