Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanedioic acid, 1,10-dibutyl ester | 109-43-3 | 98.0% | 314.47 | 10 G
SDP

Catalog No. 5000454558
Encompass_Preferred

Dibutyl sebacate (Dibutyl decanedioate) is a dibutyl ester of sebacic acid, primarily utilized as a plasticizer in the production of plastics. It is intended for research use only.

  • Utilized as a plasticizer in the production of plastics.
  • Purity 98.0%.
  • Molecular weight 314.47.
  • Store at room temperature for 3 years.
  • In solvent, store at -80°C for 2 years or -20°C for 1 year.
  • Appearance is a colorless to light yellow liquid.

Catalog No. 50-004-54558 Supplier Medchemexpress LLC Supplier No. HYW01380710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibutyl sebacate (Dibutyl decanedioate) is a dibutyl ester of sebacic acid, primarily utilized as a plasticizer in the production of plastics. It is intended for research use only.

  • Utilized as a plasticizer in the production of plastics.
  • Purity 98.0%.
  • Molecular weight 314.47.
  • Store at room temperature for 3 years.
  • In solvent, store at -80°C for 2 years or -20°C for 1 year.
  • Appearance is a colorless to light yellow liquid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.