Learn More
Medchemexpress LLC HY-W014787 100mg Medchemexpress, Decanedioic acid CAS:111-20-6 Purity:>98%
SDP
Catalog No. 501970702
Encompass_Preferred
Medchemexpress, HY-W014787 100mg Decanedioic acid CAS:111-20-6 Decanedioic acid, a normal urinary acid, is found to be associated with carnitine-acylcarnitine translocase deficiency and medium chain acyl-CoA dehydrogenase deficiency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0702 Supplier Medchemexpress LLC Supplier No. HYW014787100MG
Medchemexpress, HY-W014787 100mg Decanedioic acid CAS:111-20-6 Decanedioic acid, a normal urinary acid, is found to be associated with carnitine-acylcarnitine translocase deficiency and medium chain acyl-CoA dehydrogenase deficiency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.