Learn More
Medchemexpress LLC Decanoic acid-13C | 84600-66-8 | MFCD00083932 | 98.0% | 173.26 g/mol | C9[13C]H20O2 | 100 MG
SDP
Catalog No. 5000280704
Encompass_Preferred
Decanoic acid-13C is a carbon-13 isotopically labeled decanoic (capric) acid intended for use as an analytical standard, tracer, or internal standard in biochemical and mass spectrometry applications. The label is located at the carbonyl (1-position). The material is supplied as a small research pack for laboratory use and is provided with datasheet, COA, and SDS documentation.
  • Isotopically labeled at the carbonyl (1-13C) for tracer and metabolic studies.
  • High purity (98.0%) suitable for analytical workflows.
  • Small 100 mg package size for bench-scale experiments.
  • Useful as an internal standard in mass spectrometry and quantitative assays.
  • Chemical formula C9[13C]H20O2 and molecular weight 173.26 g/mol for sample calculations.
Catalog No. 50-002-80704 Supplier Medchemexpress LLC Supplier No. HYW266188100MG
Decanoic acid-13C is a carbon-13 isotopically labeled decanoic (capric) acid intended for use as an analytical standard, tracer, or internal standard in biochemical and mass spectrometry applications. The label is located at the carbonyl (1-position). The material is supplied as a small research pack for laboratory use and is provided with datasheet, COA, and SDS documentation.
  • Isotopically labeled at the carbonyl (1-13C) for tracer and metabolic studies.
  • High purity (98.0%) suitable for analytical workflows.
  • Small 100 mg package size for bench-scale experiments.
  • Useful as an internal standard in mass spectrometry and quantitative assays.
  • Chemical formula C9[13C]H20O2 and molecular weight 173.26 g/mol for sample calculations.