Learn More
Medchemexpress LLC Decanoic acid, 2-octyl- | 619-39-6 | MFCD00506623 | 99.9% | 284.48 g/mol | C18H36O2 | 250 MG
SDP
Catalog No. 5000281770
Encompass_Preferred
2-Octyldecanoic acid is a saturated C18 fatty acid used as a research reagent in lipid chemistry and related laboratory applications. It contains a carboxylic acid functional group and a long aliphatic chain. The compound is well characterized by CAS number 619-39-6, molecular formula C18H36O2, and molecular weight 284.48 g/mol.
  • High purity (99.93% by HPLC).
  • Identified by CAS number 619-39-6.
  • Molecular formula C18H36O2 and molecular weight 284.48 g/mol.
  • Supplied with a certificate of analysis.
  • Suitable for research use in lipid chemistry and synthesis.
  • Store and handle according to the certificate of analysis.
Catalog No. 50-002-81770 Supplier Medchemexpress LLC Supplier No. HYW176477250MG
2-Octyldecanoic acid is a saturated C18 fatty acid used as a research reagent in lipid chemistry and related laboratory applications. It contains a carboxylic acid functional group and a long aliphatic chain. The compound is well characterized by CAS number 619-39-6, molecular formula C18H36O2, and molecular weight 284.48 g/mol.
  • High purity (99.93% by HPLC).
  • Identified by CAS number 619-39-6.
  • Molecular formula C18H36O2 and molecular weight 284.48 g/mol.
  • Supplied with a certificate of analysis.
  • Suitable for research use in lipid chemistry and synthesis.
  • Store and handle according to the certificate of analysis.