Learn More
Medchemexpress LLC Decanoic acid | 334-48-5 | MFCD00004441 | 172.27 g/mol | C10H20O2 | 1 ML
SDP
Catalog No. 5000303739
Encompass_Preferred
Decanoic acid is a medium-chain fatty acid provided as a 10 mM solution in DMSO (1 mL) for research and analytical applications. It is a brain-penetrant, non-competitive AMPA receptor inhibitor with reported effects on mTORC1 signaling, tyrosinase activity, melanosome maturation, and c-Met-related pathways, making it useful for neuroscience, epilepsy models, and cell-signaling studies.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • Brain-penetrant modulator of AMPA receptor activity.
  • Reported to modulate mTORC1 signaling and c-Met pathways.
  • Reduces tyrosinase activity and inhibits melanosome maturation.
  • Suitable for neuroscience, cell-signaling, and analytical applications.
Catalog No. 50-003-03739 Supplier Medchemexpress LLC Supplier No. HYW01530910MM/1ML
Decanoic acid is a medium-chain fatty acid provided as a 10 mM solution in DMSO (1 mL) for research and analytical applications. It is a brain-penetrant, non-competitive AMPA receptor inhibitor with reported effects on mTORC1 signaling, tyrosinase activity, melanosome maturation, and c-Met-related pathways, making it useful for neuroscience, epilepsy models, and cell-signaling studies.
  • Ready-to-use 10 mM solution in DMSO (1 mL).
  • Brain-penetrant modulator of AMPA receptor activity.
  • Reported to modulate mTORC1 signaling and c-Met pathways.
  • Reduces tyrosinase activity and inhibits melanosome maturation.
  • Suitable for neuroscience, cell-signaling, and analytical applications.