Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decanoic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013950 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3950 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3950 Supplier MilliporeSigma Supelco Supplier No. 214095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decanoic acid (DA) is a saturated medium-chain (10 carbons) fatty acid primarily used in the manufacture of plastics, cosmetics, and lubricants. DA is naturally found in coconut and palm oil.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 27°C to 32°C
Boiling Point 268°C to 270°C
Density 0.893 g/mL (at 25°C)
CAS 334-48-5
Quantity 5 g
Flash Point 110°C
Physical Form Neat
Linear Formula CH3(CH2)8 COOH
MDL Number MFCD00004441
Vapor Pressure 15 mmHg (160°C)
Synonym 1-Nonanecarboxylic acid; Acid C10; C10:0; Capric acid; NSC 5025
Formula Weight 172.26
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.