Learn More
Medchemexpress LLC Decanoic acid (capric acid) | 334-48-5 | MFCD00004441 | ≥98.0% | 172.27 g/mol | C10H20O2 | 500 MG
SDP
Catalog No. 5000302593
Encompass_Preferred
Decanoic acid (capric acid) is a medium-chain saturated fatty acid used as a research reagent. It is reported to be brain-penetrant and acts as a non-competitive AMPA receptor inhibitor with antiseizure activity in animal models; it has also been studied for tyrosinase inhibition and effects on mTORC1 signaling and hepatocellular carcinoma.
  • Medium-chain saturated fatty acid with formula C10H20O2 and MW 172.27 g/mol.
  • Reported brain-penetrant, non-competitive AMPA receptor inhibitor.
  • Used in preclinical antiseizure and epilepsy research.
  • Demonstrated tyrosinase inhibition and impact on melanosome maturation.
  • Relevant for studies of mTORC1 signaling and cancer-related pathways.
  • Provided with NMR-based purity confirmation (≥98.0%).
Catalog No. 50-003-02593 Supplier Medchemexpress LLC Supplier No. HYW015309500MG
Decanoic acid (capric acid) is a medium-chain saturated fatty acid used as a research reagent. It is reported to be brain-penetrant and acts as a non-competitive AMPA receptor inhibitor with antiseizure activity in animal models; it has also been studied for tyrosinase inhibition and effects on mTORC1 signaling and hepatocellular carcinoma.
  • Medium-chain saturated fatty acid with formula C10H20O2 and MW 172.27 g/mol.
  • Reported brain-penetrant, non-competitive AMPA receptor inhibitor.
  • Used in preclinical antiseizure and epilepsy research.
  • Demonstrated tyrosinase inhibition and impact on melanosome maturation.
  • Relevant for studies of mTORC1 signaling and cancer-related pathways.
  • Provided with NMR-based purity confirmation (≥98.0%).