Learn More
Medchemexpress LLC Decanoic acid-d19 | 88170-22-3 | 98.4% | 191.38 | C10HD19O2 | 10 MG
SDP
Catalog No. 5000302577
Encompass_Preferred
Decanoic acid-d19 is the deuterium-labeled analogue of decanoic (capric) acid provided as a stable-isotope reagent for analytical and metabolic studies. It preserves the fatty acid backbone while incorporating 19 deuterium atoms, allowing clear mass and NMR separation from unlabeled material. Typical purity is 98.4% and the formula is C10HD19O2.
  • Deuterium-labeled (d19) fatty acid for isotope tracing.
  • High purity suitable for analytical applications (98.4%).
  • Useful as an internal standard for GC-MS and LC-MS quantitation.
  • Compatible with NMR and metabolic labeling studies.
  • Available in multiple small package sizes for research use.
  • COA and SDS available to support quality and handling.
Catalog No. 50-003-02577 Supplier Medchemexpress LLC Supplier No. HYW015309S110MG
Decanoic acid-d19 is the deuterium-labeled analogue of decanoic (capric) acid provided as a stable-isotope reagent for analytical and metabolic studies. It preserves the fatty acid backbone while incorporating 19 deuterium atoms, allowing clear mass and NMR separation from unlabeled material. Typical purity is 98.4% and the formula is C10HD19O2.
  • Deuterium-labeled (d19) fatty acid for isotope tracing.
  • High purity suitable for analytical applications (98.4%).
  • Useful as an internal standard for GC-MS and LC-MS quantitation.
  • Compatible with NMR and metabolic labeling studies.
  • Available in multiple small package sizes for research use.
  • COA and SDS available to support quality and handling.