Learn More
Medchemexpress LLC Decanoic acid-d19 | 88170-22-3 | 98.4% | 191.38 g/mol | C10HD19O2 | 50 MG
SDP
Catalog No. 5000302620
Encompass_Preferred
Decanoic acid-d19 is the deuterium-labeled form of decanoic (capric) acid (C10HD19O2, MW 191.38 g/mol). It is supplied as a high-purity, isotope-labeled reagent for analytical and research applications including mass spectrometry, metabolic tracing, and method development.
  • Deuterium-labeled decanoic acid for stable isotope experiments.
  • High isotopic enrichment and purity (98.4%).
  • Suitable as an internal standard for mass spectrometry.
  • Provided with certificate of analysis and safety data for traceability.
  • Available in small pack sizes for analytical workflows.
Catalog No. 50-003-02620 Supplier Medchemexpress LLC Supplier No. HYW015309S150MG
Add to Cart
Add to Cart
Decanoic acid-d19 is the deuterium-labeled form of decanoic (capric) acid (C10HD19O2, MW 191.38 g/mol). It is supplied as a high-purity, isotope-labeled reagent for analytical and research applications including mass spectrometry, metabolic tracing, and method development.
  • Deuterium-labeled decanoic acid for stable isotope experiments.
  • High isotopic enrichment and purity (98.4%).
  • Suitable as an internal standard for mass spectrometry.
  • Provided with certificate of analysis and safety data for traceability.
  • Available in small pack sizes for analytical workflows.