Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanoic acid-d3 | 102611-15-4 | MFCD00144050 | 99.0% | 175.28 | C10H17D3O2 | 5 MG
SDP

Catalog No. 5000302625
Encompass_Preferred

Decanoic acid-d3 is the deuterium-labeled form of decanoic (capric) acid used as a stable isotope reference and internal standard for analytical and research applications. It is suitable for quantitative mass spectrometry, tracer experiments, and method development, and is supplied with supporting quality and safety documentation.

  • Deuterium-labeled internal standard for GC-/LC-MS quantification.
  • Purity ≥99.0%.
  • Molecular formula C10H17D3O2; molecular weight 175.28.
  • Available in small milligram quantities for analytical use.
  • Supplied with certificate of analysis and safety data sheet.
  • Suitable for tracer and metabolic studies.

Catalog No. 50-003-02625 Supplier Medchemexpress LLC Supplier No. HYW015309S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decanoic acid-d3 is the deuterium-labeled form of decanoic (capric) acid used as a stable isotope reference and internal standard for analytical and research applications. It is suitable for quantitative mass spectrometry, tracer experiments, and method development, and is supplied with supporting quality and safety documentation.

  • Deuterium-labeled internal standard for GC-/LC-MS quantification.
  • Purity ≥99.0%.
  • Molecular formula C10H17D3O2; molecular weight 175.28.
  • Available in small milligram quantities for analytical use.
  • Supplied with certificate of analysis and safety data sheet.
  • Suitable for tracer and metabolic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.