Learn More
Medchemexpress LLC Decanoic acid-d3 | 102611-15-4 | MFCD00144050 | ≥99.0% | 175.28 g·mol⁻¹ | C10H17D3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW015309S10MG
Encompass_Preferred
Decanoic acid-d3 is the deuterated analogue of decanoic (capric) acid supplied as a stable isotope reagent for analytical and biochemical research. It is provided as a solid with high isotopic and chemical purity and is intended for laboratory research use only (not for human or clinical use).
  • Deuterated internal standard for mass spectrometry and tracer studies.
  • High isotopic and chemical purity (≥99.0%).
  • Suitable for quantitative analytical assays and method development.
  • Supplied as a solid in mg-scale quantities for precise weighing.
  • Store sealed, away from moisture and light; in solution, store at -80°C for long-term stability.
Catalog No. 50-003-02752