Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanoic acid-d3 | 102611-15-4 | MFCD00144050 | ≥99.0% | 175.28 g/mol | C10H17D3O2 | 100 MG
SDP

Catalog No. 5000303736
Encompass_Preferred

Decanoic acid-d3 is a deuterium-labeled analogue of decanoic (capric) acid used as an isotopically labeled internal standard and tracer in analytical workflows. It supports accurate quantitation in NMR, GC-MS, and LC-MS and is supplied as a white to off-white solid.

  • CAS number: 102611-15-4.
  • Purity: ≥99.0%.
  • Molecular formula: C10H17D3O2; molecular weight: 175.28 g/mol.
  • Application: internal standard and tracer for NMR, GC-MS, or LC-MS quantitative analysis.
  • Appearance: white to off-white solid.
  • Storage: sealed, away from moisture and light; in solvent, -80°C for 6 months or -20°C for 1 month.
  • Solubility: ethanol and DMF ≥25 mg/mL; DMSO ≥10 mg/mL.

Catalog No. 50-003-03736 Supplier Medchemexpress LLC Supplier No. HYW015309S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decanoic acid-d3 is a deuterium-labeled analogue of decanoic (capric) acid used as an isotopically labeled internal standard and tracer in analytical workflows. It supports accurate quantitation in NMR, GC-MS, and LC-MS and is supplied as a white to off-white solid.

  • CAS number: 102611-15-4.
  • Purity: ≥99.0%.
  • Molecular formula: C10H17D3O2; molecular weight: 175.28 g/mol.
  • Application: internal standard and tracer for NMR, GC-MS, or LC-MS quantitative analysis.
  • Appearance: white to off-white solid.
  • Storage: sealed, away from moisture and light; in solvent, -80°C for 6 months or -20°C for 1 month.
  • Solubility: ethanol and DMF ≥25 mg/mL; DMSO ≥10 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.