Learn More
Medchemexpress LLC Decanoic acid-d3 | 102611-15-4 | MFCD00144050 | 99.0% | 175.28 g·mol⁻¹ | C10H17D3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW015309S50MG
Encompass_Preferred
Decanoic acid-d3 is the deuterium-labeled analogue of decanoic (capric) acid, supplied as a white to off-white solid for use as an analytical internal standard, tracer, or research reagent. It provides high isotopic enrichment for accurate quantitation in GC-MS and LC-MS workflows and is intended for laboratory research use.
  • Deuterium-labeled internal standard for accurate quantitation.
  • Approximately 99.0% isotopic enrichment for reliable tracer studies.
  • White to off-white solid suitable for standard storage and handling.
  • Compatible with GC-MS and LC-MS analytical methods.
  • Useful as a tracer in metabolic and lipid research.
Catalog No. 50-003-02927