Learn More
Medchemexpress LLC Decanoic acid-d5 | 1219803-00-5 | 98.3% | 177.30 | C10H15D5O2 | 25 MG
SDP
Catalog No. 5000303386
Encompass_Preferred
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic (capric) acid provided for research use. It is used as an isotopically labeled standard or tracer in analytical workflows, particularly mass spectrometry and metabolic studies, and is supplied with confirmed chemical and isotopic purity.
  • Isotopically labeled (d5) decanoic acid for analytical standards.
  • High chemical and isotopic purity for accurate quantitation.
  • Available in multiple pre-weighed sizes to suit experimental needs.
  • Suitable as an internal standard or tracer in mass spectrometry.
  • Provided for research use only; not for human or diagnostic applications.
Catalog No. 50-003-03386 Supplier Medchemexpress LLC Supplier No. 25MGHYW015309S3
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic (capric) acid provided for research use. It is used as an isotopically labeled standard or tracer in analytical workflows, particularly mass spectrometry and metabolic studies, and is supplied with confirmed chemical and isotopic purity.
  • Isotopically labeled (d5) decanoic acid for analytical standards.
  • High chemical and isotopic purity for accurate quantitation.
  • Available in multiple pre-weighed sizes to suit experimental needs.
  • Suitable as an internal standard or tracer in mass spectrometry.
  • Provided for research use only; not for human or diagnostic applications.