Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanoic acid-d5 | 1219803-00-5 | 98.3% | 177.30 g/mol | C10H15D5O2 | 5 MG
SDP

Catalog No. 5000303674
Encompass_Preferred

Decanoic acid-d5 is the deuterium-labeled analogue of decanoic acid provided for research applications including analytical tracing and pharmacology studies. It is supplied as a small oil sample with high isotopic purity suitable for use in formulation and bioanalytical workflows.

  • Deuterium-labeled decanoic acid for tracer and analytical studies.
  • High isotopic and chemical purity suitable for research applications.
  • Reported purity: 98.3%.
  • Molecular weight approximately 177.30 g/mol; formula C10H15D5O2.
  • Appearance: colorless to off-white oil.
  • DMSO soluble (≈200 mg/mL) with recommended handling notes for hygroscopic solvent.
  • Available in a 5 mg pack size for small-scale experiments.

Catalog No. 50-003-03674 Supplier Medchemexpress LLC Supplier No. HYW015309S35MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decanoic acid-d5 is the deuterium-labeled analogue of decanoic acid provided for research applications including analytical tracing and pharmacology studies. It is supplied as a small oil sample with high isotopic purity suitable for use in formulation and bioanalytical workflows.

  • Deuterium-labeled decanoic acid for tracer and analytical studies.
  • High isotopic and chemical purity suitable for research applications.
  • Reported purity: 98.3%.
  • Molecular weight approximately 177.30 g/mol; formula C10H15D5O2.
  • Appearance: colorless to off-white oil.
  • DMSO soluble (≈200 mg/mL) with recommended handling notes for hygroscopic solvent.
  • Available in a 5 mg pack size for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.