Learn More
Medchemexpress LLC Decanoic acid-d5 | 1219803-00-5 | 98.3% | 177.30 g/mol | C10H15D5O2 | 50 MG
SDP
Catalog No. 5000303673
Encompass_Preferred
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic acid used as a stable isotope reagent for analytical and research applications. It is suited for tracer experiments, method development, and quantitative analyses requiring labeled internal standards.
  • Deuterium-labeled analogue for tracer and mechanistic studies.
  • High chemical purity (98.3%).
  • Available in multiple small-scale sizes suitable for analytical workflows.
  • Compatible with mass spectrometry and nuclear magnetic resonance methods.
  • Useful as an internal standard for quantitative assays and metabolic studies.
Catalog No. 50-003-03673 Supplier Medchemexpress LLC Supplier No. HYW015309S350MG
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic acid used as a stable isotope reagent for analytical and research applications. It is suited for tracer experiments, method development, and quantitative analyses requiring labeled internal standards.
  • Deuterium-labeled analogue for tracer and mechanistic studies.
  • High chemical purity (98.3%).
  • Available in multiple small-scale sizes suitable for analytical workflows.
  • Compatible with mass spectrometry and nuclear magnetic resonance methods.
  • Useful as an internal standard for quantitative assays and metabolic studies.