Learn More
Medchemexpress LLC Decanoic acid-d5 | 1219803-00-5 | MFCD01317497 | 98.3% | 177.30 | C10H15D5O2 | 10 MG
SDP
Catalog No. 5000302505
Encompass_Preferred
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic acid, provided as a high-purity stable isotope reagent for analytical and biological research. It serves as an internal standard, tracer, or reference compound in mass spectrometry and metabolic studies, enabling confident identification and quantification through a predictable mass shift.
  • High isotopic labeling (D5) for clear mass separation.
  • High purity (98.3%) suitable for analytical workflows.
  • Compact research-scale quantity for easy storage and handling.
  • Soluble in DMSO at high concentration for convenient stock solutions.
  • Stable under recommended storage conditions for long-term use.
  • Suitable as an internal standard in mass spectrometry and tracer studies.
Catalog No. 50-003-02505 Supplier Medchemexpress LLC Supplier No. HYW015309S310MG
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic acid, provided as a high-purity stable isotope reagent for analytical and biological research. It serves as an internal standard, tracer, or reference compound in mass spectrometry and metabolic studies, enabling confident identification and quantification through a predictable mass shift.
  • High isotopic labeling (D5) for clear mass separation.
  • High purity (98.3%) suitable for analytical workflows.
  • Compact research-scale quantity for easy storage and handling.
  • Soluble in DMSO at high concentration for convenient stock solutions.
  • Stable under recommended storage conditions for long-term use.
  • Suitable as an internal standard in mass spectrometry and tracer studies.