Learn More
Medchemexpress LLC Decanoic acid, ethyl ester | 110-38-3 | MFCD00009581 | 99.3% | 200.32 g/mol | C12H24O2 | 100 G
SDP
Catalog No. 5000303907
Encompass_Preferred
Ethyl decanoate (decanoic acid, ethyl ester) is a medium-chain fatty acid ester supplied for research and industrial applications. It is commonly used in flavor and fragrance formulations and as an intermediate in organic synthesis. The material is offered as a high-purity reagent characterized by a molecular weight of 200.32 g/mol and a GC purity of about 99.3%.
  • High purity suitable for research applications.
  • Commonly used as a flavoring and fragrance component.
  • Serves as an intermediate in organic synthesis.
  • Characterized by molecular formula C12H24O2 and molecular weight 200.32 g/mol.
  • Available in multiple pack sizes including 100 g.
Catalog No. 50-003-03907 Supplier Medchemexpress LLC Supplier No. HYW014884100G
Ethyl decanoate (decanoic acid, ethyl ester) is a medium-chain fatty acid ester supplied for research and industrial applications. It is commonly used in flavor and fragrance formulations and as an intermediate in organic synthesis. The material is offered as a high-purity reagent characterized by a molecular weight of 200.32 g/mol and a GC purity of about 99.3%.
  • High purity suitable for research applications.
  • Commonly used as a flavoring and fragrance component.
  • Serves as an intermediate in organic synthesis.
  • Characterized by molecular formula C12H24O2 and molecular weight 200.32 g/mol.
  • Available in multiple pack sizes including 100 g.