Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanoic acid, ethyl ester | 110-38-3 | MFCD00009581 | 99.3% | 200.32 g/mol | C12H24O2 | 25 G
SDP

Catalog No. 5000303560
Encompass_Preferred

Ethyl decanoate (decanoic acid ethyl ester) is a high-purity fatty acid ethyl ester supplied as a colorless to light yellow liquid. It is commonly used as a reagent and as an intermediate or ingredient in flavor and fragrance formulations, and is suitable for analytical and biochemical applications requiring a well-characterized ester.

  • Purity 99.30% (GC).
  • Molecular formula C12H24O2.
  • Molecular weight 200.32 g/mol.
  • Physical form: liquid; density 0.862 g/cm3.
  • Pack size available: 25 g (also 50 g, 100 g).
  • Storage: store at room temperature, shelf life 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Typical uses: flavor and fragrance formulation, analytical standard, biochemical reagent.

Catalog No. 50-003-03560 Supplier Medchemexpress LLC Supplier No. 25GHYW014884
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl decanoate (decanoic acid ethyl ester) is a high-purity fatty acid ethyl ester supplied as a colorless to light yellow liquid. It is commonly used as a reagent and as an intermediate or ingredient in flavor and fragrance formulations, and is suitable for analytical and biochemical applications requiring a well-characterized ester.

  • Purity 99.30% (GC).
  • Molecular formula C12H24O2.
  • Molecular weight 200.32 g/mol.
  • Physical form: liquid; density 0.862 g/cm3.
  • Pack size available: 25 g (also 50 g, 100 g).
  • Storage: store at room temperature, shelf life 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Typical uses: flavor and fragrance formulation, analytical standard, biochemical reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.