Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanoic acid, ethyl ester | 110-38-3 | MFCD00009581 | 99.3% | 200.32 g/mol | C12H24O2 | 50 G
SDP

Catalog No. 5000303274
Encompass_Preferred

Ethyl decanoate (decanoic acid ethyl ester) is a colorless to light-yellow liquid ester used as a flavoring agent and a research reagent. Supplied at high purity, it is suitable for analytical work, formulation, and flavor and fragrance studies requiring a stable medium-chain fatty acid ethyl ester.

  • High purity, suitable for analytical use.
  • Liquid form; colorless to light yellow for easy identification.
  • Molecular weight 200.32 g/mol and chemical formula C12H24O2.
  • Density 0.862 g/cm³ for formulation calculations.
  • Available in laboratory pack sizes for research workflows.
  • COA and SDS available for quality and safety documentation.
  • Store at room temperature; follow solvent storage recommendations for extended stability.

Catalog No. 50-003-03274 Supplier Medchemexpress LLC Supplier No. HYW01488450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethyl decanoate (decanoic acid ethyl ester) is a colorless to light-yellow liquid ester used as a flavoring agent and a research reagent. Supplied at high purity, it is suitable for analytical work, formulation, and flavor and fragrance studies requiring a stable medium-chain fatty acid ethyl ester.

  • High purity, suitable for analytical use.
  • Liquid form; colorless to light yellow for easy identification.
  • Molecular weight 200.32 g/mol and chemical formula C12H24O2.
  • Density 0.862 g/cm³ for formulation calculations.
  • Available in laboratory pack sizes for research workflows.
  • COA and SDS available for quality and safety documentation.
  • Store at room temperature; follow solvent storage recommendations for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.