Learn More
Medchemexpress LLC Decanoic acid sodium 25g | 1002-62-6 | 194.25 g/mol | C10H19NaO2 | 25 G
SDP
Catalog No. 5000307506
Encompass_Preferred
Decanoic acid, sodium salt (sodium decanoate) is the sodium salt of decanoic (capric) acid supplied as a white to off-white powder. It is water-soluble and commonly used as a biochemical reagent and formulation excipient where it acts as a penetration enhancer to increase drug absorption; it also has antibacterial properties useful in preservative and cosmetic applications.
  • High purity (manufacturer-reported) suitable for research and formulation use.
  • Water-soluble sodium salt for easy incorporation into aqueous systems.
  • Acts as a penetration enhancer to improve absorption across biological membranes.
  • Exhibits antibacterial properties useful for preservative applications.
  • Available in small pack sizes for laboratory research.
Catalog No. 50-003-07506 Supplier Medchemexpress LLC Supplier No. HY11534025G
Decanoic acid, sodium salt (sodium decanoate) is the sodium salt of decanoic (capric) acid supplied as a white to off-white powder. It is water-soluble and commonly used as a biochemical reagent and formulation excipient where it acts as a penetration enhancer to increase drug absorption; it also has antibacterial properties useful in preservative and cosmetic applications.
  • High purity (manufacturer-reported) suitable for research and formulation use.
  • Water-soluble sodium salt for easy incorporation into aqueous systems.
  • Acts as a penetration enhancer to improve absorption across biological membranes.
  • Exhibits antibacterial properties useful for preservative applications.
  • Available in small pack sizes for laboratory research.