Learn More
Medchemexpress LLC Decanoic acid sodium 5g | 1002-62-6 | 5 G
SDP
Catalog No. 5000227094
Encompass_Preferred
Decanoic acid sodium (sodium decanoate) is the sodium salt of the medium-chain fatty acid decanoic (capric) acid. It is a white to off-white, water-soluble solid used in biochemical and pharmaceutical research as a penetration enhancer and for formulation testing where antimicrobial properties are relevant.
  • High purity (99.92%).
  • Molecular formula C10H19NaO2 and molecular weight 194.25.
  • White to off-white solid, water soluble.
  • Available in small pack sizes suitable for laboratory use (5 g, 25 g, 50 g).
  • Used as a penetration enhancer to increase absorption across biological membranes.
  • Exhibits antimicrobial activity useful for preservative and formulation studies.
Catalog No. 50-002-27094 Supplier Medchemexpress LLC Supplier No. HY1153405G
Decanoic acid sodium (sodium decanoate) is the sodium salt of the medium-chain fatty acid decanoic (capric) acid. It is a white to off-white, water-soluble solid used in biochemical and pharmaceutical research as a penetration enhancer and for formulation testing where antimicrobial properties are relevant.
  • High purity (99.92%).
  • Molecular formula C10H19NaO2 and molecular weight 194.25.
  • White to off-white solid, water soluble.
  • Available in small pack sizes suitable for laboratory use (5 g, 25 g, 50 g).
  • Used as a penetration enhancer to increase absorption across biological membranes.
  • Exhibits antimicrobial activity useful for preservative and formulation studies.