Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decanophenone, 98+%

Catalog No. AAL0692314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0692314 25 g
AAL0692306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0692314 Supplier Thermo Scientific Chemicals Supplier No. L0692314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6048-82-4 | C16H24O | 232.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6048-82-4
Molecular Formula C16H24O
Molecular Weight (g/mol) 232.37
MDL Number MFCD00009582
InChI Key QQXJNLYVPPBERR-UHFFFAOYSA-N
Synonym decanophenone, 1-decanone, 1-phenyl, n-decanophenone, capriphenone, ketone, nonyl phenyl, ketone, nonenyl phenyl, nonyl phenyl ketone, 1-phenyl-1-decanone, acmc-209mkb
PubChem CID 80148
IUPAC Name 1-phenyldecan-1-one
SMILES CCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 34°C to 36°C
Boiling Point 165°C to 167°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 2048790
Formula Weight 232.37
Percent Purity ≥98%
Chemical Name or Material Decanophenone
EINECSNumber 227-946-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.