Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Decanoyl chloride

Catalog No. 140295500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-029-5500ML 500 mL
14-029-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-029-5500ML Supplier Sigma Aldrich Supplier No. 140295500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Decanoyl chloride enhances the activity of proleather from bacillus sp
  • During the synthesis of poly(lactic acid) in organic solvents.decanoyl chloride was used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units

Specifications

CAS 112-13-0
Density 0.919 g/mL (at 25°C (literature))
Boiling Point 94°C to 96°C (5 mmHg)
Molecular Formula C10H19ClO
Refractive Index n20/D 1.441 (literature)
Linear Formula CH3(CH2)8 COCl
MDL Number MFCD00000771
Quantity 500 mL
Synonym Capric acid chloride
Molecular Weight (g/mol) 190.71
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.