Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Decanoyl coenzyme A | 1264-57-9 | MFCD00079131 | 99.9% | 921.78 g·mol⁻¹ | C31H54N7O17P3S | 5 MG
SDP

Catalog No. 5000223408
Encompass_Preferred

Decanoyl coenzyme A is an acyl-CoA derivative used as a primer for fatty acid elongation in Mycobacterium smegmatis and as a biochemical reagent in metabolic and enzymatic assays. It is provided as a high-purity research compound suitable for use as a reference standard or substrate in enzymology and lipid metabolism studies.

  • Primer for fatty acid elongation assays.
  • High purity: 99.9%.
  • Molecular formula C31H54N7O17P3S and molecular weight 921.78 g·mol⁻¹.
  • Storage: store at -20°C, protect from light; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
  • Intended for research use only.

Catalog No. 50-002-23408 Supplier Medchemexpress LLC Supplier No. HY1378685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Decanoyl coenzyme A is an acyl-CoA derivative used as a primer for fatty acid elongation in Mycobacterium smegmatis and as a biochemical reagent in metabolic and enzymatic assays. It is provided as a high-purity research compound suitable for use as a reference standard or substrate in enzymology and lipid metabolism studies.

  • Primer for fatty acid elongation assays.
  • High purity: 99.9%.
  • Molecular formula C31H54N7O17P3S and molecular weight 921.78 g·mol⁻¹.
  • Storage: store at -20°C, protect from light; in solvent store at -80°C up to 6 months or -20°C up to 1 month.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.