Learn More
Medchemexpress LLC Decanoyl-RVKR-CMK | 150113-99-8 | MFCD00798792 | 98.0% | 744.41 Da | C34H66ClN11O5 | 5 MG
SDP
Catalog No. 5000240783
Encompass_Preferred
Decanoyl-RVKR-CMK is a peptide-based proprotein convertase inhibitor used in biochemical and cellular research. It inhibits furin and related convertases, blocking envelope glycoprotein precursor processing and reducing viral replication in cell models. Supplied as a solid for small-scale laboratory studies.
  • Peptide-based inhibitor of furin and proprotein convertases.
  • Blocks envelope glycoprotein precursor processing in cells.
  • Reduces viral replication in cellular assays.
  • Suitable for in vitro biochemical and cell-based studies.
  • Provided as a solid for convenient handling and storage.
Catalog No. 50-002-40783 Supplier Medchemexpress LLC Supplier No. HY1077605MG
Add to Cart
Add to Cart
Decanoyl-RVKR-CMK is a peptide-based proprotein convertase inhibitor used in biochemical and cellular research. It inhibits furin and related convertases, blocking envelope glycoprotein precursor processing and reducing viral replication in cell models. Supplied as a solid for small-scale laboratory studies.
  • Peptide-based inhibitor of furin and proprotein convertases.
  • Blocks envelope glycoprotein precursor processing in cells.
  • Reduces viral replication in cellular assays.
  • Suitable for in vitro biochemical and cell-based studies.
  • Provided as a solid for convenient handling and storage.