Learn More
Medchemexpress LLC Decanoyl-RVKR-CMK | 150113-99-8 | MFCD00798792 | 98.0% | 744.41 g·mol⁻1 | C34H66ClN11O5 | 1 MG
SDP
Catalog No. 5000240897
Encompass_Preferred
Decanoyl-RVKR-CMK is a peptidyl chloromethyl ketone inhibitor of subtilisin/Kex2p-like proprotein convertases (including furin). It is used experimentally to block proprotein convertase activity, inhibit gp160 processing, and reduce viral replication in cell-based and biochemical assays.
  • Peptidyl chloromethyl ketone inhibitor of proprotein convertases.
  • Inhibits gp160 processing and viral replication in experimental systems.
  • Suitable for cell-based and biochemical assays.
  • White to off-white solid supplied in small mass packages.
  • Reported purity about 98.0% and molecular weight 744.41 g·mol⁻1.
  • Used to study furin-dependent protein processing and viral entry.
Catalog No. 50-002-40897 Supplier Medchemexpress LLC Supplier No. HY1077601MG
Decanoyl-RVKR-CMK is a peptidyl chloromethyl ketone inhibitor of subtilisin/Kex2p-like proprotein convertases (including furin). It is used experimentally to block proprotein convertase activity, inhibit gp160 processing, and reduce viral replication in cell-based and biochemical assays.
  • Peptidyl chloromethyl ketone inhibitor of proprotein convertases.
  • Inhibits gp160 processing and viral replication in experimental systems.
  • Suitable for cell-based and biochemical assays.
  • White to off-white solid supplied in small mass packages.
  • Reported purity about 98.0% and molecular weight 744.41 g·mol⁻1.
  • Used to study furin-dependent protein processing and viral entry.