Learn More
Medchemexpress LLC Decanoyl-RVKR-CMK trifluoroacetate | 2098497-25-5 | 97.1% | 858.43 g·mol⁻¹ | C36H67ClF3N11O7 | 1 MG
SDP
Catalog No. 5000240784
Encompass_Preferred
Decanoyl-RVKR-CMK trifluoroacetate is a synthetic peptide inhibitor of subtilisin/kexin-like proprotein convertases, including furin. It blocks processing of viral envelope glycoprotein precursors (gp160) and reduces HIV replication in cell-based assays. The material is supplied as the trifluoroacetate salt and is intended for research use.
Catalog No. 50-002-40784 Supplier Medchemexpress LLC Supplier No. HY107760A1MG
Decanoyl-RVKR-CMK trifluoroacetate is a synthetic peptide inhibitor of subtilisin/kexin-like proprotein convertases, including furin. It blocks processing of viral envelope glycoprotein precursors (gp160) and reduces HIV replication in cell-based assays. The material is supplied as the trifluoroacetate salt and is intended for research use.