Learn More
Apexbio Technology LLC (±)-Decanoylcarnitine chloride 14919-36-9 50mg
SDP
Catalog No. 5000090857
Encompass_Preferred
racemic Baclofen -Decanoylcarnitine chloride is a small-molecule modulator targeting cholinergic systems by engaging acetylcholine receptors including nicotinic and muscarinic subtypes It is designed to influence neurotransmission and lipid metabolism racemic Baclofen -Decanoylcarnitine chloride exerts its biological activity primarily through modulation of acetylcholine receptors and inhibition of -aminobutyric acid (GABA) receptor activity thereby attenuating GABA-mediated inhibitory synaptic currents In neuronal cells and cardiac models such acylcarnitine derivatives demonstrate antiarrhythmic properties post-ischemic insult and modulation of pain signaling involving the phospholipase C-inositol triphosphate pathway Based on these pharmacological properties racemic Baclofen -Decanoylcarnitine chloride holds research potential in cardiac arrhythmia and pain modulation applications
Catalog No. 50-000-90857 Supplier Apexbio Technology LLC Supplier No. B631550
racemic Baclofen -Decanoylcarnitine chloride is a small-molecule modulator targeting cholinergic systems by engaging acetylcholine receptors including nicotinic and muscarinic subtypes It is designed to influence neurotransmission and lipid metabolism racemic Baclofen -Decanoylcarnitine chloride exerts its biological activity primarily through modulation of acetylcholine receptors and inhibition of -aminobutyric acid (GABA) receptor activity thereby attenuating GABA-mediated inhibitory synaptic currents In neuronal cells and cardiac models such acylcarnitine derivatives demonstrate antiarrhythmic properties post-ischemic insult and modulation of pain signaling involving the phospholipase C-inositol triphosphate pathway Based on these pharmacological properties racemic Baclofen -Decanoylcarnitine chloride holds research potential in cardiac arrhythmia and pain modulation applications