Learn More
Integra Biosciences Corp Decanting tubing MC 30
SDP
Catalog No. 5000881961
Encompass
Decanting tubing MEDIACLAVE 30 complete - for insertion into the sterilization chamber/cuvette for dispensing incl rigid stainless steel tube (length 613mm) silicone tubing securing nut
Catalog No. 50-008-81961 Supplier Integra Biosciences Corp Supplier No. 136061
Decanting tubing MEDIACLAVE 30 complete - for insertion into the sterilization chamber/cuvette for dispensing incl rigid stainless steel tube (length 613mm) silicone tubing securing nut