Learn More
Medchemexpress LLC Decapeptide-12 | 137665-91-9 | 99.95% | 1395.52 | 5 MG
SDP
Catalog No. 5000425892
Encompass_Preferred
Decapeptide-12 is a small oligopeptide that acts as a tyrosinase inhibitor, interacting with the C-terminal residue of tyrosinase. It is a competitive inhibitor of mushroom tyrosinase and increases the transcription of SIRT, reducing melanin content in melanocytes. It is utilized in research concerning melanogenesis, senescence, and inflammation.
  • Acts as a tyrosinase inhibitor
  • Reduces melanin content in melanocytes
  • Increases transcription of SIRT
  • Used for research in melanogenesis, senescence, and inflammation
  • High purity of 99.95%
Catalog No. 50-004-25892 Supplier Medchemexpress LLC Supplier No. HYP00965MG
Decapeptide-12 is a small oligopeptide that acts as a tyrosinase inhibitor, interacting with the C-terminal residue of tyrosinase. It is a competitive inhibitor of mushroom tyrosinase and increases the transcription of SIRT, reducing melanin content in melanocytes. It is utilized in research concerning melanogenesis, senescence, and inflammation.
  • Acts as a tyrosinase inhibitor
  • Reduces melanin content in melanocytes
  • Increases transcription of SIRT
  • Used for research in melanogenesis, senescence, and inflammation
  • High purity of 99.95%