Learn More
Braintree Scientific DecapiCones, for Mice, 2 Dispensers, 200 cones total
SDP
Catalog No. NC9679094
Encompass
Make injections and decapitation quicker and easier with Braintree Scientific's DecapiCones. Tapered plastic film tubes provide quick and easy restraint of rats, mice, and other small animals. I.P. injections can be made directly through the film! DecapiCones restrain post-decapitation kicking and prevent personal contact with feces or urine. A unique dispenser holds DecapiCones open and ready for use. Simply hold the DecapiCone in one hand and introduce the animal with the other. Animals enter readily, heading for the breathing hole at the small end. Roll and squeeze the large end closed. They may be used repeatedly for injections and simply discarded when soiled. For decapitation, hold at the rear and insert the small end into the decapitator. **Sold for research purposes only and not for human use**
Catalog No. NC9679094 Supplier Braintree Scientific Supplier No. MDC200
Make injections and decapitation quicker and easier with Braintree Scientific's DecapiCones. Tapered plastic film tubes provide quick and easy restraint of rats, mice, and other small animals. I.P. injections can be made directly through the film! DecapiCones restrain post-decapitation kicking and prevent personal contact with feces or urine. A unique dispenser holds DecapiCones open and ready for use. Simply hold the DecapiCone in one hand and introduce the animal with the other. Animals enter readily, heading for the breathing hole at the small end. Roll and squeeze the large end closed. They may be used repeatedly for injections and simply discarded when soiled. For decapitation, hold at the rear and insert the small end into the decapitator. **Sold for research purposes only and not for human use**