Learn More
Poltex Decapper Tool Holder - 4 Position
SDP
Catalog No. 01259211
Change view
Click to view available options
Mounting Type:
Magnet Mount
Tape Mount
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
01-259-211 Magnet Mount
01-259-200 Tape Mount
01-259-145 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
N/A
Catalog No. 01-259-211 Supplier Poltex Supplier No. DECAPR4M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A
Holds 4 screw cap decapper tools. Choose from Wall Mount, HB Tape Mount, or Magnet Mount with durable and cleanable PETG construction.
  • Holds 4 Thermo Fisher screw cap decapper tools
  • Available in Wall Mount (screws and anchors included)
  • Available in HB Tape Mount (high-bond tape included)
  • Available in Magnet Mount (powerful magnets included)
  • Durable and cleanable PETG construction
  • Consumables not included
Specifications
Quantity 1 Ea.
Mounting Type Magnet Mount
For Use With (Application) Holds 4 Thermofisher Screw Cap Decapper Tools
Includes Magnets for Mounting
Dimensions (L x W x H) 4.8 x 10.8 x 4.7 in.
For Use With (Equipment) Thermofisher Screw Cap Decapper Tools
Material PETG