Learn More
AdipoGen Decarestrictine D
SDP
Catalog No. 502562018
Encompass_Preferred
Chemical. CAS 127393-89-9. Formula C10H16O5. MW 216.2. Isolated from Penicillium sp. Hypolipidemic. Cholesterol biosynthesis inhibitor.
Catalog No. 50-256-2018 Supplier AdipoGen Supplier No. BVT0283M001
Chemical. CAS 127393-89-9. Formula C10H16O5. MW 216.2. Isolated from Penicillium sp. Hypolipidemic. Cholesterol biosynthesis inhibitor.