Learn More
Cayman Chemical Decatromicin B, Purity: ≥95%, CAS: 235097-64-0, 2.5 mg
SDP
Catalog No. 502940855
Encompass_Preferred
A bacterial metabolite; active against Gram-positive bacteria, including several strains of S. aureus (MICs = 0.39-0.78 µg/ml), two strains of MRSA (MICs = 0.39 and 0.78 µg/ml), as well as M. luteus, B. subtilis, and C. bovis (MICs = 0.78, 0.78, and 6.25 µg/ml, respectively)
Catalog No. 50-294-0855 Supplier Cayman Chemical Supplier No. 281322.5 MG
A bacterial metabolite; active against Gram-positive bacteria, including several strains of S. aureus (MICs = 0.39-0.78 µg/ml), two strains of MRSA (MICs = 0.39 and 0.78 µg/ml), as well as M. luteus, B. subtilis, and C. bovis (MICs = 0.78, 0.78, and 6.25 µg/ml, respectively)