Learn More
TARGETMOL CHEMICALS INC DECERNOTINIB 10MG
SDP
Catalog No. 502577074
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Decernotinib (VRT-831509)(VX-509; VRT-831509) is a potent and selective Janus kinase 3 (JAK3) inhibitor with Ki of 2.5 nM; IC50 is 50-170 nM in cellular assays. purity: 99%
Catalog No. 50-257-7074 Supplier TARGETMOL CHEMICALS INC Supplier No. T263610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Decernotinib (VRT-831509)(VX-509; VRT-831509) is a potent and selective Janus kinase 3 (JAK3) inhibitor with Ki of 2.5 nM; IC50 is 50-170 nM in cellular assays. purity: 99%