Learn More
Selleck Chemical LLC Decernotinib (VX-509) 2mg 944842-54-0
SDP
Catalog No. 501364922
Encompass_Preferred
Decernotinib (VX-509) is a potent and selective JAK3 inhibitor with Ki of 2.5 nM, >4-fold selectivity over JAK1, JAK2, and TYK2, respectively. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4922 Supplier Selleck Chemical LLC Supplier No. S75412MG
Decernotinib (VX-509) is a potent and selective JAK3 inhibitor with Ki of 2.5 nM, >4-fold selectivity over JAK1, JAK2, and TYK2, respectively. Phase 2/3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.