Learn More
Cambridge Isotope Laboratories Dechlorane 604 (component A) (unlabeled) 100 ug/mL in nonane 1 2 mL
Catalog No. 5000168767
Encompass
Dechlorane 604 (component A) (unlabeled) 100 ug/mL in nonane 1 2 mL
Catalog No. 50-001-68767 Supplier Cambridge Isotope Laboratories Supplier No. ULM96221.2
Dechlorane 604 (component A) (unlabeled) 100 ug/mL in nonane 1 2 mL