Learn More
Medchemexpress LLC HY-A0004 100mg Medchemexpress, Decitabine CAS:2353-33-5 Purity:>98%
SDP
Catalog No. 502029373
Encompass_Preferred
Medchemexpress, HY-A0004 100mg Decitabine CAS:2353-33-5 Decitabine (NSC 127716) is a deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9373 Supplier Medchemexpress LLC Supplier No. HYA0004100MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-A0004 100mg Decitabine CAS:2353-33-5 Decitabine (NSC 127716) is a deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.