Learn More
Selleck Chemical LLC Decitabine 10mM/1mL 2353-33-5 Deoxycytidine, Dacogen, 5-aza-2'-deoxycytidine, 5-AZA-dC, 5-aza-CdR
SDP
Catalog No. NC0881140
Encompass_Preferred
Decitabine (Deoxycytidine, Dacogen, 5-aza-2-deoxycytidine, 5-AZA-dC, 5-aza-CdR) is a DNA methyltransferase inhibitor, incorporating into DNA and resulting in hypomethylation of DNA and intra-S-phase arrest of DNA replication. It is used to treat myelodysplastic syndrome (MDS). Decitabine induces cell cycle arrest and apoptosis in various cancer cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0881140 Supplier Selleck Chemical LLC Supplier No. S120010MM/1ML
Decitabine (Deoxycytidine, Dacogen, 5-aza-2-deoxycytidine, 5-AZA-dC, 5-aza-CdR) is a DNA methyltransferase inhibitor, incorporating into DNA and resulting in hypomethylation of DNA and intra-S-phase arrest of DNA replication. It is used to treat myelodysplastic syndrome (MDS). Decitabine induces cell cycle arrest and apoptosis in various cancer cell lines. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.