Learn More
Medchemexpress LLC HY-A0004 500mg , Decitabine CAS:2353-33-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0004500MG
Encompass_Preferred
HY-A0004 500mg Decitabine CAS: 2353-33-5 Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1102