Learn More
Medchemexpress LLC Decitabine (Standard) | 2353-33-5 | 99.9% | C8H12N4O4 | 5 MG
SDP
Catalog No. 5000433072
Encompass_Preferred
Decitabine (Standard) is the analytical standard of Decitabine, intended for research and analytical applications. It is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. It incorporates into DNA in place of cytosine, covalently trapping DNA methyltransferase and causing irreversible enzyme inhibition, leading to cell G2/M arrest and apoptosis. This compound demonstrates potent anticancer activity.
  • Analytical standard for Decitabine
  • Intended for research and analytical applications
  • Orally active deoxycytidine analogue antimetabolite
  • DNA methyltransferase inhibitor
  • Induces cell G2/M arrest and apoptosis
  • Demonstrates potent anticancer activity
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-33072 Supplier Medchemexpress LLC Supplier No. HYA0004R5MG
Decitabine (Standard) is the analytical standard of Decitabine, intended for research and analytical applications. It is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. It incorporates into DNA in place of cytosine, covalently trapping DNA methyltransferase and causing irreversible enzyme inhibition, leading to cell G2/M arrest and apoptosis. This compound demonstrates potent anticancer activity.
  • Analytical standard for Decitabine
  • Intended for research and analytical applications
  • Orally active deoxycytidine analogue antimetabolite
  • DNA methyltransferase inhibitor
  • Induces cell G2/M arrest and apoptosis
  • Demonstrates potent anticancer activity
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS