Learn More
MSC Vikan Deck Scrub, 16 inch, Stiff, Yellow
Catalog No. 502838842
Encompass
This deck scrub is designed for effective scrubbing of surfaces with stubborn messes. It features an angled arrangement of Bristle Security Units for efficient cleaning and lacks resin, glue, and staples, making the bristle units easy to clean, FDA-compliant, and more secure.
  • Made from FDA compliant raw materials.
  • Autoclave safe up to 250°F.
  • Dishwasher safe.
  • Stiff bristles.
  • Hygienic Euro thread.
  • UST Hygienic design.
  • Unique bristle pattern.
Catalog No. 50-283-8842 Supplier MSC Supplier No. 54807235
This deck scrub is designed for effective scrubbing of surfaces with stubborn messes. It features an angled arrangement of Bristle Security Units for efficient cleaning and lacks resin, glue, and staples, making the bristle units easy to clean, FDA-compliant, and more secure.
  • Made from FDA compliant raw materials.
  • Autoclave safe up to 250°F.
  • Dishwasher safe.
  • Stiff bristles.
  • Hygienic Euro thread.
  • UST Hygienic design.
  • Unique bristle pattern.