Learn More
MSC Vikan Deck Scrub Brush, Stiff Polyester, 15.7 inch, White
Catalog No. 502838843
Encompass
This deck scrub brush is designed for effective scrubbing of stubborn dirt on both textured and smooth surfaces. It features Ultra Safe Technology (UST) with enhanced bristle security units directly molded into the brush block, ensuring strong bristle retention and eliminating contamination risks.
  • Enhanced bristle security units are directly molded into the brush block, providing unprecedented bristle retention strength.
  • Eliminates the risk of contamination associated with traditional resin set or staple set brushes.
  • Very effective in scrubbing stubborn dirt on both textured and smooth surfaces.
  • Helps prevent cross-contamination by ensuring the brush is used only in its designated area.
  • Individually molded bristles further reduce contamination risks.
Catalog No. 50-283-8843 Supplier MSC Supplier No. 54807243
This deck scrub brush is designed for effective scrubbing of stubborn dirt on both textured and smooth surfaces. It features Ultra Safe Technology (UST) with enhanced bristle security units directly molded into the brush block, ensuring strong bristle retention and eliminating contamination risks.
  • Enhanced bristle security units are directly molded into the brush block, providing unprecedented bristle retention strength.
  • Eliminates the risk of contamination associated with traditional resin set or staple set brushes.
  • Very effective in scrubbing stubborn dirt on both textured and smooth surfaces.
  • Helps prevent cross-contamination by ensuring the brush is used only in its designated area.
  • Individually molded bristles further reduce contamination risks.