Learn More
Medchemexpress LLC Decloxizine dihydrochloride | 13073-96-6 | 99.69% | C21H30Cl2N2O2 | 25 MG
SDP
Catalog No. 5000357633
Encompass_Preferred
Decloxizine dihydrochloride is a histamine 1 receptor antagonist.
  • H1 receptor antagonist
  • Suitable for neurological research
  • Applicable in studies of eye or ear disease
  • Relevant for metabolic or endocrine disease research
  • Used in inflammation or immune system disease studies
  • Involved in neurodegenerative disease research
  • Related to lipid metabolism
  • Applicable to Parkinson's disease research
  • Relevant for obesity studies
  • Used in endocrinology research
Catalog No. 50-003-57633 Supplier Medchemexpress LLC Supplier No. 25MGHYA0075
Decloxizine dihydrochloride is a histamine 1 receptor antagonist.
  • H1 receptor antagonist
  • Suitable for neurological research
  • Applicable in studies of eye or ear disease
  • Relevant for metabolic or endocrine disease research
  • Used in inflammation or immune system disease studies
  • Involved in neurodegenerative disease research
  • Related to lipid metabolism
  • Applicable to Parkinson's disease research
  • Relevant for obesity studies
  • Used in endocrinology research