Learn More
Medchemexpress LLC Decloxizine dihydrochloride | 13073-96-6 | 99.7% | C21H30Cl2N2O2 | 500 MG

Supplier: Medchemexpress LLC HYA0075500MG
Decloxizine dihydrochloride acts as a histamine 1 receptor antagonist and is intended for research use only. This compound presents as a white to off-white solid with a high purity of 99.69%.
- Antagonizes histamine 1 receptors
- Solid form
- High purity of 99.69%
- Suitable for various research applications
- Recommended storage at 4°C in sealed conditions, away from moisture
- Solutions can be stored at -80°C for up to 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.