Learn More
Cayman Chemical Decanoic Acid-d19, Purity: ≥99% deuterated forms (d1-d19), CAS: 88170-22-3, 100 mg
SDP
Catalog No. 502940342
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-294-0342 Supplier Cayman Chemical Supplier No. 28871100 MG
An internal standard for the quantification of decanoic acid by GC- or LC-MS