Learn More
Cayman Chemical Decanoic Acid-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 62716-49-8, 100 mg
SDP
Catalog No. 502988313
Encompass_Preferred
An internal standard for the quantification of decanoic acid by GC- or LC-MS
Catalog No. 50-298-8313 Supplier Cayman Chemical Supplier No. 29409100 MG
An internal standard for the quantification of decanoic acid by GC- or LC-MS